Learn More
Hach Company Contact Conduct Sens-3/4in NPT
Catalog No. 5000207514 Shop All Hach Company Products
Encompass
General Purpose Contacting Conductivity Sensor-3/4 inch male NPT Threaded PPS Body-30 ft CableNominal Cell Constant (K) 0 5 3/4 inch male NPT threaded PPS body 9m (30ft) Analog Cable Graphite Electrode
Catalog No. 50-002-07514 Supplier Hach Company Supplier No. 3433B8A30N
General Purpose Contacting Conductivity Sensor-3/4 inch male NPT Threaded PPS Body-30 ft CableNominal Cell Constant (K) 0 5 3/4 inch male NPT threaded PPS body 9m (30ft) Analog Cable Graphite Electrode