Learn More
Hach Company Contact Conduct Sens-PPS-Low
Catalog No. 5000207686 Shop All Hach Company Products
Encompass
General Purpose Contacting Conductivity Sensor-PPS Body-Low Conductivity (k 0 5)Contacting conductivity sensor with a nominal cell constant k 0 5 cm-1 -inch male NPT threaded PPS body 6 m analogue cable and graphite electrode Measuring range 0 - 1000 S/cm 0001 - 20 M
Catalog No. 50-002-07686 Supplier Hach Company Supplier No. 3433B8A
General Purpose Contacting Conductivity Sensor-PPS Body-Low Conductivity (k 0 5)Contacting conductivity sensor with a nominal cell constant k 0 5 cm-1 -inch male NPT threaded PPS body 6 m analogue cable and graphite electrode Measuring range 0 - 1000 S/cm 0001 - 20 M