Learn More
Azzota Corp Contact lens refract no ATC
Catalog No. 5000887285
Encompass_Preferred
Contact Lens Tester Refractometer Water range 35-80% Accuracy /- 1% Without ATC
Catalog No. 50-008-87285 Supplier Azzota Corp Supplier No. RHL3580
Contact Lens Tester Refractometer Water range 35-80% Accuracy /- 1% Without ATC