Learn More
Electron Microscopy Sciences Contact Plate, convex bottom, 60x15 mm, 15 ml
SDP
Catalog No. 501900275
Encompass_Preferred
These polystyrene dishes are free from optical distortion. The dish has 16 of 10 x 10 mm square grids, which is marked numerically in one direction and alphabetically in the other. Dishes are packaged 20 per sleeve and are radiation sterile. Convex bottom saves on culture medium.
Catalog No. 50-190-0275 Supplier Electron Microscopy Sciences Supplier No. 7069220
These polystyrene dishes are free from optical distortion. The dish has 16 of 10 x 10 mm square grids, which is marked numerically in one direction and alphabetically in the other. Dishes are packaged 20 per sleeve and are radiation sterile. Convex bottom saves on culture medium.