Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ Contact Plates
SCI_ED

Catalog No. S012400 Shop All Fischer Technical Company Products
Change view
Click to view available options
Media Type:
Levine EMB Agar
Potato Dextrose Agar
SDA with Lecithin and Tween 80
TSA w/ Lecithin and Tween 80
Tryptic Soy Agar
Detects:
Bacteria
Bacteria and Fungi
Fungi
Gram Negative Bacteria
Microorganisms
Catalog No. Media Type Detects
S012400 Tryptic Soy Agar Bacteria
S012393 SDA with Lecithin and Tween 80 Fungi
S012395 TSA w/ Lecithin and Tween 80 Microorganisms
S012396 Potato Dextrose Agar Bacteria and Fungi
S012398 Levine EMB Agar Gram Negative Bacteria
5 options
Prop 65: Cancer and Reproductive Harm.

Catalog No. S012400

Supplier: Fischer Technical Company™ CM100RP15

Only null left
Add to Cart
Add to Cart
Prop 65: Cancer and Reproductive Harm.

Contact plates ideal for testing surfaces.

  • Available in a variety of formulations
  • For environmental monitoring of surfaces
  • Steam sterilized
  • Agar is slightly raised to make it easy to test surfaces

Specifications

Detects Bacteria
Format Contact Plate
Manufacturing Origin US with Globally Sourced Materials
Media Type Tryptic Soy Agar
Product Type Contact Plate
Sterility Sterile
Target Organism Class Fastidious and Non-fastidious Organisms
Test Type Microbiological Tests
For Use With (Application) Microbiological Testing
Quantity 10 Plates
Dimensions (Dia. x H) 65 x 15 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.