Learn More
Medchemexpress LLC Contactin-3/CNTN3 M 50ug
SDP
Catalog No. 5000178523
Encompass_Preferred
Contactin-3/CNTN3 protein facilitates cell surface interactions in nervous system development promoting neurite outgrowth and interacting with PTPRG to regulate neuronal growth and connectivity Contactin-3/CNTN3 Protein Mouse (HEK293 His) is the recombinant mouse-derived Contactin-3/CNTN3 protein expressed by HEK293 with C-His labeled tag
Catalog No. 50-001-78523 Supplier Medchemexpress LLC Supplier No. HYP7422850UG
Contactin-3/CNTN3 protein facilitates cell surface interactions in nervous system development promoting neurite outgrowth and interacting with PTPRG to regulate neuronal growth and connectivity Contactin-3/CNTN3 Protein Mouse (HEK293 His) is the recombinant mouse-derived Contactin-3/CNTN3 protein expressed by HEK293 with C-His labeled tag