Learn More
AdipoGen Contactin-6 hFc h rec
SDP
Catalog No. 502562967
Encompass_Preferred
Protein. Human contactin-6 aa 1-999 is fused at the C-terminus to the Fc portion of human IgG. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. Contactin-6 is a member of the contactins, which mediate cell surface interactions during nervous system development. Contactin-6 has a key role in the formation of glutamatergic, but not GABAergic, synapses during postnatal development of the hippocampal formation as well as the cerebellum. It is involved in oligodendrocytes generation by acting as a ligand of Notch1. It is also involved in motor coordination.
Catalog No. 50-256-2967 Supplier AdipoGen Supplier No. AG40A0156C010
Protein. Human contactin-6 aa 1-999 is fused at the C-terminus to the Fc portion of human IgG. Source HEK 293 cells. Endotoxin content <0.1EU/µg purified protein LAL test Lonza. Liquid. 0.2µm-filtered solution in PBS. Purity >90% SDS-PAGE. Contactin-6 is a member of the contactins, which mediate cell surface interactions during nervous system development. Contactin-6 has a key role in the formation of glutamatergic, but not GABAergic, synapses during postnatal development of the hippocampal formation as well as the cerebellum. It is involved in oligodendrocytes generation by acting as a ligand of Notch1. It is also involved in motor coordination.