Learn More
ACROBiosystems Recombinant Protein;Human Contactin-2 / Tag1 / CNTN2 Protein;HEK293;100UG;CN2,CNTN2,CNTN-2,AXT,DKFZp781D102,FLJ37193,FLJ42746,MGC157722,TAG-1,TAG1,TAX,TAX1,TAX-1,Contactin-2,Contactin 2
Catalog No. 501657522
Encompass_Preferred
Recombinant Protein;Human Contactin-2 / Tag1 / CNTN2 Protein;100UG;Human Contactin-2, His Tag (CN2-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 31 - Asn 1012 (Accession # NP_005067.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 109 kDa. The protein migrates as 140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7522 Supplier ACROBiosystems Supplier No. CN2H5226100UG
Recombinant Protein;Human Contactin-2 / Tag1 / CNTN2 Protein;100UG;Human Contactin-2, His Tag (CN2-H5226) is expressed from human 293 cells (HEK293). It contains AA Ser 31 - Asn 1012 (Accession # NP_005067.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 109 kDa. The protein migrates as 140 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.