Learn More
Hach Company Contacting conductivity sensor, K = 0.5 cm⁻¹, 3/4 in NPT PVDF compression fitting, 20 ft (6 m) cable, titanium electrodes, built-in Pt1000 RTD
Catalog No. 5000207617 Shop All Hach Company Products
Encompass
Contacting conductivity sensor with a 0.5 cm⁻¹ cell constant, 3/4 in NPT PVDF compression fitting, 6 m (20 ft) cable, titanium electrodes, and a built-in Pt1000 RTD temperature element. Designed for precise conductivity and resistivity measurements from ultra-pure water to high conductivity process streams, it provides fast temperature compensation and simple integration with digital SC controllers.
  • Built-in Pt1000 RTD provides fast, accurate temperature compensation.
  • Titanium electrodes offer corrosion resistance for extended service life.
  • PVDF compression fitting enables straightforward installation and up to 4 in insertion depth.
  • 0.5 cm⁻¹ cell constant suitable for low-conductivity measurements.
  • 6 m (20 ft) cable allows flexible mounting and routing.
  • Measures a wide conductivity range from ultra-pure to high conductivity fluids.
  • Plug-and-play digital compatibility for simplified setup and operation.
Catalog No. 50-002-07617 Supplier Hach Company Supplier No. 3422B3A20G
Contacting conductivity sensor with a 0.5 cm⁻¹ cell constant, 3/4 in NPT PVDF compression fitting, 6 m (20 ft) cable, titanium electrodes, and a built-in Pt1000 RTD temperature element. Designed for precise conductivity and resistivity measurements from ultra-pure water to high conductivity process streams, it provides fast temperature compensation and simple integration with digital SC controllers.
  • Built-in Pt1000 RTD provides fast, accurate temperature compensation.
  • Titanium electrodes offer corrosion resistance for extended service life.
  • PVDF compression fitting enables straightforward installation and up to 4 in insertion depth.
  • 0.5 cm⁻¹ cell constant suitable for low-conductivity measurements.
  • 6 m (20 ft) cable allows flexible mounting and routing.
  • Measures a wide conductivity range from ultra-pure to high conductivity fluids.
  • Plug-and-play digital compatibility for simplified setup and operation.