Learn More
Cole-Parmer Cole-Parmer Collapsible Container, Low-Density Polyethylene (LDPE), 2.5 gallon; 10/Pk
Catalog No. NC9799403 Shop All Cole-Parmer Products
Encompass_Preferred
These containers nest uniformly when empty and expand readily when filled with liquid. They include polypropylene (PP) caps with injection-molded threads, replacement caps sold separately.
Catalog No. NC9799403 Supplier Cole-Parmer Supplier No. 0610030
These containers nest uniformly when empty and expand readily when filled with liquid. They include polypropylene (PP) caps with injection-molded threads, replacement caps sold separately.