Learn More
Cole-Parmer Cole-Parmer Collapsible Container, Low-Density Polyethylene (LDPE), 1 gallon; 12/Pk
Catalog No. NC9222101 Shop All Cole-Parmer Products
Encompass_Preferred
These containers nest uniformly when empty and expand readily when filled with liquid. They include polypropylene (PP) caps with injection-molded threads, replacement caps sold separately.
Catalog No. NC9222101 Supplier Cole-Parmer Supplier No. 0610020
These containers nest uniformly when empty and expand readily when filled with liquid. They include polypropylene (PP) caps with injection-molded threads, replacement caps sold separately.