Learn More
Sklar Instruments Basket, Sklar Sterilization Mid Size Container Basket, Sklar 16 in. x 10 in. x 4 in. EA.
SDP
Catalog No. 501182926 Shop All Sklar Instruments Products
Encompass_Preferred
Basket, SklarLite Sterilization Mid Size Container Basket, Sklar, 16 in. x 10 in. x 4 in. SklarLite Sterilization Mid Size Container Wire Basket, SklarLite Sterilization Container Baskets are made from anodized aluminum that allow them to be durable but light. Sklar offers a wide range of accessories, including baskets, silicone mats, custom ID labels, and filters. This product is a mid size wire basket designed to insert clamps to stabilize instruments during sterilization.
Catalog No. 50-118-2926 Supplier Sklar Instruments Supplier No. 104484
Basket, SklarLite Sterilization Mid Size Container Basket, Sklar, 16 in. x 10 in. x 4 in. SklarLite Sterilization Mid Size Container Wire Basket, SklarLite Sterilization Container Baskets are made from anodized aluminum that allow them to be durable but light. Sklar offers a wide range of accessories, including baskets, silicone mats, custom ID labels, and filters. This product is a mid size wire basket designed to insert clamps to stabilize instruments during sterilization.