Learn More
Med Vet International Biohazard Disposal Bags, 2-4 Gallon, J1067, 50-Pack
Catalog No. 5000495701
Encompass_Preferred
The days have passed when veterinarians simply disposed of their hazardous waste like surgical gloves, drape, and animal body parts in the trash. Medical standards require responsible disposal of medical waste in appropriately marked bags that can be safely autoclaved. Orange-red with a large biohazard symbol and precautionary in four languages are clearly visible for safety. Made of high molecular weight, high density (HMHD/polyethylene) and are steam autoclavable up to 121°C (250°F).
Catalog No. 50-004-95701 Supplier Med Vet International Supplier No. JOR1067
The days have passed when veterinarians simply disposed of their hazardous waste like surgical gloves, drape, and animal body parts in the trash. Medical standards require responsible disposal of medical waste in appropriately marked bags that can be safely autoclaved. Orange-red with a large biohazard symbol and precautionary in four languages are clearly visible for safety. Made of high molecular weight, high density (HMHD/polyethylene) and are steam autoclavable up to 121°C (250°F).