Learn More
Sklar Instruments Container, Sklar Sterilization Half Size Container Bottom, 11 3/4 in. x 11 in. x 8 in. Sklar Sterilization Half Size Container Bottom Non-Perforated Bottom, Silver. EA.
SDP
Catalog No. 501182949 Shop All Sklar Instruments Products
Encompass_Preferred
Container, SklarLite Sterilization Half Size Container Bottom, 11 3/4 in. x 11 in. x 8 in. SklarLite Sterilization Half Size Container Bottom Non-Perforated Bottom, Silver, SklarLite Sterilization Containers are made from anodized aluminum that allow them to be durable but lighter than traditional containers, ideal for not only sterilization and storage, but also easy transport. Available in multiple sizes, with lids available in a variety of colors, containers are available in sets or individual components. Sklar also offers a wide range of accessories, including baskets, silicone mats, custom ID labels, and filters. This product is a half size non-perforated bottom in silver color.
Catalog No. 50-118-2949 Supplier Sklar Instruments Supplier No. 104746
Container, SklarLite Sterilization Half Size Container Bottom, 11 3/4 in. x 11 in. x 8 in. SklarLite Sterilization Half Size Container Bottom Non-Perforated Bottom, Silver, SklarLite Sterilization Containers are made from anodized aluminum that allow them to be durable but lighter than traditional containers, ideal for not only sterilization and storage, but also easy transport. Available in multiple sizes, with lids available in a variety of colors, containers are available in sets or individual components. Sklar also offers a wide range of accessories, including baskets, silicone mats, custom ID labels, and filters. This product is a half size non-perforated bottom in silver color.