Learn More
Andwin Scientific Sharps Container 1gal CS40
SDP
Catalog No. NC1619851
Encompass_Preferred
Sharps Disposal Container Ideal for lancets scalpels slides pipets and syringes. Locking rotor secures lid. Round with red lid. Volume: 4.7L. Dimensions: 16.7Lx22Diameter cm.
This item is not returnable. View return policy
Catalog No. NC1619851 Supplier Andwin Scientific Supplier No. 15704196EA
Sharps Disposal Container Ideal for lancets scalpels slides pipets and syringes. Locking rotor secures lid. Round with red lid. Volume: 4.7L. Dimensions: 16.7Lx22Diameter cm.