Learn More
BURKLE INC Container screw joint with lock nut R2", PP, Ø 2.36 in. (60 mm)
Catalog No. 5000025712
Encompass_Preferred
Also for wide-necked screw caps. The screw joint 2" is screwed on from the inside using lock nut 2". (Mounting holes in the container 60 mm).
The container connectors make decanting even easier and safer with their more secure connection to barrels. The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange.
Catalog No. 50-000-25712 Supplier BURKLE INC Supplier No. 56003140
Also for wide-necked screw caps. The screw joint 2" is screwed on from the inside using lock nut 2". (Mounting holes in the container 60 mm).
The container connectors make decanting even easier and safer with their more secure connection to barrels. The connectors secure the pump in the barrel, enable a vertical position change and allow the required air exchange.