Learn More
MSC Globe Scientific Specimen Container, 500 mL, Polypropylene, Non-Sterile, 100/Pk.
Catalog No. 5000516599
Encompass
These 500 mL (16 oz.) specimen containers are designed for reliable storage and transport of laboratory samples, particularly in histology applications. Each container is constructed from durable polypropylene and includes a separate yellow screw cap made of polyethylene for a secure seal.
  • Capacity of 500 mL (16 oz.)
  • Constructed from polypropylene
  • Includes separate yellow polyethylene screw cap
  • Ribbed cap design allows for easy opening and closing
  • Features molded graduations
  • Stackable design for efficient storage
  • Non-sterile
  • Case of 100 containers
Catalog No. 50-005-16599 Supplier MSC Supplier No. 09450313
These 500 mL (16 oz.) specimen containers are designed for reliable storage and transport of laboratory samples, particularly in histology applications. Each container is constructed from durable polypropylene and includes a separate yellow screw cap made of polyethylene for a secure seal.
  • Capacity of 500 mL (16 oz.)
  • Constructed from polypropylene
  • Includes separate yellow polyethylene screw cap
  • Ribbed cap design allows for easy opening and closing
  • Features molded graduations
  • Stackable design for efficient storage
  • Non-sterile
  • Case of 100 containers