Learn More
Med Vet International Sharps Container, 1 Gallon, each
Catalog No. 502522183
Encompass_Preferred
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy-to-assemble. Meets all EPA and OSHA standards. Nestable containers save space and reduce shipping costs.
Catalog No. 50-252-2183 Supplier Med Vet International Supplier No. SHARP1G
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy-to-assemble. Meets all EPA and OSHA standards. Nestable containers save space and reduce shipping costs.