Learn More
Med Vet International Sharps Container, 1 Quart, 3/Pack
Catalog No. 502522181
Encompass_Preferred
Portable sharps containers are designed to fit into blood drawing trays, also provides point-of-use disposal and features a square base for added stability. Choice of single or dual openings accommodates a variety of sharps. Containers lock for final disposal and safe transport
Catalog No. 50-252-2181 Supplier Med Vet International Supplier No. SHARP1QX3
Portable sharps containers are designed to fit into blood drawing trays, also provides point-of-use disposal and features a square base for added stability. Choice of single or dual openings accommodates a variety of sharps. Containers lock for final disposal and safe transport