Learn More
Med Vet International Portable Phlebotomy Sharps Container, 1Qt
Catalog No. 502832095
Encompass_Preferred
Phlebotomy Containers: Portable sharps containers are designed to fit into blood drawing trays, also provides point-of-use disposal and features a square base for added stability. Choice of single or dual openings accommodates a variety of sharps. Containers lock for final disposal and safe transport. 1 Quart.
Catalog No. 50-283-2095 Supplier Med Vet International Supplier No. MDS705110H
Phlebotomy Containers: Portable sharps containers are designed to fit into blood drawing trays, also provides point-of-use disposal and features a square base for added stability. Choice of single or dual openings accommodates a variety of sharps. Containers lock for final disposal and safe transport. 1 Quart.