Learn More
Med Vet International Medline Multipurpose Sharps Container 2 Gallon
Catalog No. 502831769
Encompass_Preferred
These containers are designed for use in restricted access areas. Nestable containers save space and help reduce shipping costs. Non-tortuous lid design accommodates a variety of sharps sizes. Horizontal drop maximizes use of container volume. Containers lock for final disposal and safe transport. Brackets available to mount containers to wall and free up counter space
Catalog No. 50-283-1769 Supplier Med Vet International Supplier No. MDS705202H
These containers are designed for use in restricted access areas. Nestable containers save space and help reduce shipping costs. Non-tortuous lid design accommodates a variety of sharps sizes. Horizontal drop maximizes use of container volume. Containers lock for final disposal and safe transport. Brackets available to mount containers to wall and free up counter space