Learn More
Med Vet International Sharps Container, 2 Gallons, Locking Top Flap, Polypropylene, Each
Catalog No. 502522180
Encompass_Preferred
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy to assemble. Meets all EPA and OSHA standards. Nestable containers save space and reduce shipping costs. Never recycle the container. Sharps should never be thrown loosely into the trash or toilet. Even Sharps that retract after use, or are very small, should be disposed of like all other sharps. Go to Safeneedledisposal.org for more information about your states regulations. Sold in eaches.
Catalog No. 50-252-2180 Supplier Med Vet International Supplier No. SHARP2G
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy to assemble. Meets all EPA and OSHA standards. Nestable containers save space and reduce shipping costs. Never recycle the container. Sharps should never be thrown loosely into the trash or toilet. Even Sharps that retract after use, or are very small, should be disposed of like all other sharps. Go to Safeneedledisposal.org for more information about your states regulations. Sold in eaches.