Learn More
Med Vet International Sharps Container, 3 Gallon, Each
Catalog No. 502643444
Encompass_Preferred
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy to assemble. Meets all EPA and OSHA standards.
Catalog No. 50-264-3444 Supplier Med Vet International Supplier No. SHARP3G
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy to assemble. Meets all EPA and OSHA standards.