Learn More
Med Vet International Sharps Container, 4 gallon, each
Catalog No. 502522179
Encompass_Preferred
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy to assemble.
Catalog No. 50-252-2179 Supplier Med Vet International Supplier No. SHARP4G
For safe disposal of sharps in restricted access areas. Versatile lid accommodates contaminated items of various sizes. Constructed of puncture resistant polypropylene. Locking top flap. Easy to assemble.