Learn More
Electron Microscopy Sciences Clarified Polypropylene Container with lid, 180 ml
SDP
Catalog No. 501928603
Encompass_Preferred
You can have confidence that these containers are designed for the safe, leak-resistant transport and storage of fluids. Screw-top containers have knurled cap and base for easy handling. They are graduated and made of durable clarified polypropylene. Sizing include lids.
Diameter: 2-11/16 in
Height: 2-9/16 in
Catalog No. 50-192-8603 Supplier Electron Microscopy Sciences Supplier No. 6423106
Add to Cart
Add to Cart
You can have confidence that these containers are designed for the safe, leak-resistant transport and storage of fluids. Screw-top containers have knurled cap and base for easy handling. They are graduated and made of durable clarified polypropylene. Sizing include lids.
Diameter: 2-11/16 in
Height: 2-9/16 in