Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hygiena Contamination Control Kits, ATP Test
SDP

Catalog No. 14037780
Change view
Click to view available options
For Use With:
Hygiena
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With
14-037-780 Hygiena
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-037-780 Supplier Hygiena Supplier No. CCK4
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

ATP standard and 100 cuvettes

High-precision test for measuring total and free ATP in liquid samples

It provides expanded flexibility over all-in-one test devices such as Watershot™ and Aquasnap™ by allowing sample cleanup in the case of high quenching, particulates or fibers in aqueous samples. It uses unique liquid-stable reagent which eliminates reconstitution of the Contamination Control Kit comes with an ATP standard and 100 cuvettes making it a complete kit. Intended to replace the Multitrace Kit™

  • Detects microbial and nonmicrobial contamination
  • Uses liquid-stable reagents versus lyophilized - 4 bottles versus 20 vials
  • Offers flexibility when dealing with challenging samples
  • Ideal solution for measuring contamination in process water, wastewater, water tanks and cooling towers
  • Kit includes all components plus ATP standard
  • Extended shelf life - 12 months

Specifications

Content And Storage Keep refrigerated between 2° to 8°C.
For Use With Hygiena
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.