Learn More
Contec™ 2769 Series Stainless Steel Cart
BIOPHARMA
Catalog No. 19200004 Shop All Contec Inc. Products
Change view
Click to view available options
Material:
Stainless Steel
Catalog No. Material
19-200-004 Stainless Steel
1 options
Catalog No. 19-200-004 Supplier Contec™ Supplier No. 2769
Add to Cart
Add to Cart
Maintain best practice protocol, even in cleanrooms where space is an issue.The Contec™ Stainless Steel Cart is a fully autoclavable stainless steel rolling cart with two 5 gallon buckets in a compact and easily transportable size. The cart features four standard casters, as well as a stainless steel drip cap.
  • Fully autoclavable stainless-steel rolling carts with two polypropylene 5 gallon (20L) bucket (red, gray), and stainless steel drip cap
  • Durable, yet lightweight construction makes bucket systems easy to maneuver around cleanrooms
  • Cart features four standard or heavy-duty casters
  • Double bucket system allows for differentiation between cleaning and rinsing solutions
Specifications
Product Type Replacement Cart
Material Stainless Steel
Autoclavable Autoclavable
For Use With (Equipment) 20 L Bucket
Sterility Non-sterile