Learn More
Contec™ Bucket Wringers and Frames
BIOPHARMA
Catalog No. 19168797 Shop All Contec Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Contec EasyCurve Mop
Contec EasyCurve Mops, 6.5 gal. Bucket Only
Contec VertiKlean MAX or QuickTask Flat Mops, 5 gal. or 6.5 gal. Bucket
Contec VertiKlean MAX or QuickTask Flat Mops, 6.5 gal. Bucket
VertiKlean MAX and QuickTask Mops
Product Type:
Curved Mop Wringer
Flat Mop Wringer
Slinger Frame and Insert
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Product Type
19-168-797 Contec VertiKlean MAX or QuickTask Flat Mops, 6.5 gal. Bucket -
19-033-705 Contec VertiKlean MAX or QuickTask Flat Mops, 5 gal. or 6.5 gal. Bucket -
19-168-528 VertiKlean MAX and QuickTask Mops Flat Mop Wringer
19-168-529 VertiKlean MAX and QuickTask Mops Curved Mop Wringer
19-200-014 Contec EasyCurve Mop -
19-033-706 Contec EasyCurve Mops, 6.5 gal. Bucket Only Slinger Frame and Insert
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-168-797 Supplier Contec™ Supplier No. 2738
Add to Cart
Add to Cart
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
Use with Contec™ bucket systems and mops. Contec™ Bucket Wringers and Frames are available in three styles to match the selected Contec™ mop system.
  • Choose from downpress, flat or curved styles
  • Available in polypropylene or stainless steel
  • Ideal for cleanrooms and controlled environments with small autoclaves or limited storage space
  • Select wringer that complements mop
Curved stainless-steel sieve 19-200-014 for EasyCurve™ mop and flat stainless-steel sieve 19-033-715 for flat mops such as VertiKlean™ MAX and QuickTask. Sieves require stainless-steel frame 19-033-716 for 5 gal. (20 L) buckets and 19-033-717 for 6.5 gal. (25 L) buckets.
Specifications
Autoclavable Autoclavable
Disposable Reusable
For Use With (Application) Ideal for cleanrooms and controlled environments
For Use With (Equipment) Contec VertiKlean MAX or QuickTask Flat Mops, 6.5 gal. Bucket
Sterility Non-sterile