Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Edgeless™ Mop System

Catalog No. 19033960 Shop All Contec Inc. Products
Change view
Click to view available options
Head Type:
Edgeless
Enhanced with Textured Headband
Long Textured
Ribbed
Textured
Color:
Black (Tail Band)
Gray, Black (Tail Band)
White
White, Blue (Tail Band)
White, Red (Tail Band)
Quantity:
1/Bag, 20 Bags/Cs.
1/Bag, 30 Bags/Cs.
12/Cs.
20/Cs.
30/Cs.
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Head Type Color Quantity
19-033-960 Long Textured White, Blue (Tail Band) 12/Cs.
19-081-379 Edgeless White 1/Bag, 20 Bags/Cs.
19-033-850 Enhanced with Textured Headband White 20/Cs.
19-033-950 Textured White 1/Bag, 30 Bags/Cs.
19-039-940 Edgeless White 1/Bag, 30 Bags/Cs.
19-033-959 Long Textured White 30/Cs.
19-033-851 Enhanced with Textured Headband Gray, Black (Tail Band) 20/Cs.
19-033-961 Ribbed Black (Tail Band) 12/Cs.
19-033-962 Long Textured White, Red (Tail Band) 12/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. 19-033-960 Supplier Contec™ Supplier No. EL5BLU

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Clean in controlled environments and cleanrooms. The Edgeless™ Mop System is constructed from a continuous tube of 100% polyester knitted fabric. The mop heads help eliminate contamination from breakage and deterioration of traditional string mops.

  • Designed for wet mopping in controlled environments and cleanrooms
  • Ideal for use in sterile environments
  • Mop heads are constructed from a continuous tube of 100% polyester knitted fabric, looped to form strands that encapsulate liquids for maximum absorption
  • Mop heads are free of any cellulosic contaminants
  • Textured mop heads feature ribbed, twisted strands for 60% more sorbent capacity than standard Edgeless mops
  • Textured knitted polyester tube mop heads help eliminate contamination from breakage and deterioration of traditional string mops
  • Mop heads are laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom

Specifications

Product Type Mop Head
Head Type Long Textured
Color White, Blue (Tail Band)
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Limited-use
Quantity 12/Cs.
For Use With (Equipment) Contec Edgeless Mopping System
Description Cleanroom Laundered Long Textured Mop Head
For Use With (Application) Designed for use with aqueous solutions containing cleaning agents and disinfectants; compatible with wide range of chemicals
Material Polyester
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.