Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Edgeless™ Mop
BIOPHARMA

Catalog No. 19830353 Shop All Contec Inc. Products
Change view
Click to view available options
Material:
Polyester Knitted Fabric
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Material
19-830-353 Polyester Knitted Fabric
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-830-353 Supplier Contec™ Supplier No. AMGN0001
Add to Cart
Add to Cart
Request a Quote

Long textured edgeless mop

Edgeless™ Mops are made from strong, durable knitted polyester, eliminating contamination from breakage and deterioration common with cotton and rayon string mops. Cleanroom laundered Edgeless™ Mops are made from a continuous tube of 100% polyester knitted fabric looped to form mop strands that will not shed or break.

  • Durable and chemical resistant
  • Continuous loop of mop strands provides improved cleanliness, eliminating cut edges
  • Textured large head band provides scrubbing power for difficult stains and debris

Patented Textured and Long Textured Edgeless Mops feature unique ribbed, twisted strands that are more sorbent than traditional string mops. They have a textured head band that is ideal when additional abrasion resistance is required.

Specifications

Product Type Cleanroom Mop
Capacity 13 oz.
Sterility Non-sterile
Autoclavable Autoclavable
Disposable Single-use
For Use With (Application) Tough Cleaning Tasks
Color-coded No
Material Polyester Knitted Fabric
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.