Learn More
Contec™ Mop Head Saturation System
BIOPHARMAGREENER_CHOICE
Catalog No. 19168704PF Shop All Contec Inc. Products
Change view
Click to view available options
Capacity (Metric):
Not Applicable
Product Type:
Bucket
Bucket Casters
Compact Bucket System
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Product Type
19-168-704PF Not Applicable Bucket Casters
19-168-703PF - -
19-168-701PF - Compact Bucket System
19-168-702PF - Bucket
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-168-704PF Supplier Contec™ Supplier No. 2647
Add to Cart
Add to Cart
Designed to simplify the cleanroom mopping process, Contec’s Mop Head Saturation System allows mop heads to be presaturated prior to being passed into the controlled environment. This eliminates the need for any water to be passed into the cleanroom.
Small and compact, the stainless steel cart with twin 8 L buckets is ideal for small to medium sized cleanroom. Being lightweight and maneuverable the cart is easy to use. The cart is fitted with four non marking, autoclavable castors, meaning the whole cart can be autoclaved before use. In order make the system highly efficient and cost effective the mop heads used need to be fast-wicking and highly sorbent. Contec has developed a new flat mop head to be used as part of the system; Contec Quiltec I Polyester Mop Heads.
The buckets are designed to take 5 flat mop heads which can be saturated with 1 L of fluid. The second bucket can be used for the dilution of a concentrate and then to store the used mop heads preventing any cross contamination. The mop heads are placed on an easy-fit mop applicator frame which coupled with the mop head design,means the mop heads can be fitted to the mop frame hands-free. A stainless steel disinfectant diffuser tray helps to evenly saturate every mop head. The diffuser tray can be placed into a slot on the side of the cart when not in use for easy storage. Simply pour the disinfectant onto the mop heads either from the bottle or straight from the other bucket, wait 5 mins for full saturation and the mops are ready to use. Contec Quiltec I Mop Heads are manufactured from 100% 2-ply polyester around a rayon core so are ideal for even the highest grade of cleanroom as they generate very small numbers of particle and fibres.
Specifications
Product Type Bucket Casters
Capacity (Metric) Not Applicable
Material Stainless Steel
Autoclavable Autoclavable
Disposable Reusable
Includes Replacement Part, Blue Caster, Swivel with Brake Replacement