Learn More
Contec™ Caster Wheels for Carts
Catalog No. 19200001 Shop All Contec Inc. Products
Change view
Click to view available options
Material:
Polyurethane
Catalog No. Material
19-200-001 Polyurethane
1 options
Catalog No. 19-200-001 Supplier Contec™ Supplier No. 8064E
Add to Cart
Add to Cart
Replace damaged wheels on Contec™ carts for improved system use. Contec™ Caster Wheels for Carts are intended for use with Contec carts 2720 and 2721 (Cat. Nos. 19-168-744 and 19-033-778).
  • Polyurethane, autoclavable plastic
  • May be sterilized by autoclave or gamma irradiation; at 121°C for 30 minutes
Specifications
Product Type Bucket Casters
Material Polyurethane
Autoclavable Autoclavable
Sterility Non-sterile