Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
- Fully autoclavable polypropylene rolling cart with caster
- One red 6.5 gal. (25L) bucket with wringer
- Durable, lightweight construction makes system easy to maneuver around smaller cleanrooms
- For smaller cleanrooms, critical environments.
- Use with Contec Edgeless Mops
Specifications
Specifications
| Product Type | 1 Bucket System |
| Material | Polypropylene |
| Autoclavable | Autoclavable |
| Color | Red |
| For Use With (Application) | Ideal for Smaller Cleanrooms and Critical Environments |
| Includes | Red 6.5 gal. Bucket, Casters |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
