Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Contec™ Sporicidin™ Disinfectant Solution, Nonsterile

Shop All Contec Inc. Products


Click to view available options
Volume (Metric):
0.94 L
3.8 L
Description
- Provides 100% kill of pathogenic vegetative organisms, including MRSA, VRE and Avian Influenza A Virus (H9N2 and H1N1)
- Continuous residual activity up to 6 months
- Compatible with plastics, wood, glass and metals
- Non-staining, non-abrasive and non-corrosive
- Alcohol-free
- EPA and FDA registered
- OSHA compliant
- Lowest EPA toxicity category
Compliance
EPA and FDA registered, OSHA compliant, Lowest EPA toxicity category
Specifications
Specifications
| Product Type | Disinfectant Solution |
| Form | Liquid |
| pH | 7.5 to 8.5 |
| Product Line | Sporicidin™ |
| Scent | Fresh Scent |
| Sterility | Non-sterile |
| Certifications/Compliance | EPA, OSHA |
| Composition | 1.56% Buffered Phenol, 0.06% Sodium Phenate, 98.38% Inert Ingredients |
| Container Type | Trigger spray bottle |
| For Use With (Application) | Disinfecting hard surfaces in critical areas where USP <797> compliance has been mandated, such as ISO Class 8 Anterooms in hospitals and compounding pharmacies |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction