Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Sporicidin™ Enzymatic Cleaner
BIOPHARMA

Catalog No. 189991459 Shop All Contec Inc. Products
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
18-999-1459 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-999-1459 Supplier Contec™ Supplier No. ENZ1284
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Loosen and remove dried debris, protein, blood and mucin deposits. The Contec™ Sporicidin™ Enzymatic Cleaner effectively cleans both natural and synthetic fibers thats have been soiled with organic and inorganic debris. This product is ideal for household, medical, dental, institutional, healthcare, veterinary and cleanroom use,

  • Concentrated solution with a blend of enzymes for instruments and surfaces soiled with organic and inorganic debris
  • Low foaming solution formulated to loosen and remove dried debris, protein, blood and mucin deposits
  • It effectively cleans both natural and synthetic fibers that have been soiled with organic and inorganic debris
  • Sporicidin™ enzymatic cleaner can be used in ultrasonic units, automatic washers and evacuation systems
  • This highly effective cleaner and stain remover has a neutral pH and is biodegradable, nonflammable and low toxicity
  • This product is ideal for household, medical, dental, institutional, healthcare, veterinary and cleanroom use

Order Info

One gallon makes 128 ready to use gallons

Specifications

Product Type Enzymatic Cleaner
Form Liquid
Product Line Sporicidin
Composition 44.29% Enzymes, 5% surfactants, 50.71% inert ingredients
For Use With (Application) Concentrated solution with a blend of enzymes for instruments and surfaces soiled with organic and inorganic debris
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.