Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Contec™ Sporicidin™ Disinfectant Solution, Sterile
BIOPHARMA

Catalog No. 189991460 Shop All Contec Inc. Products
Change view
Click to view available options
Volume (English):
1 gal.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English)
18-999-1460 1 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-999-1460 Supplier Contec™ Supplier No. GAMMA1284F

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Disinfects, cleans, and deodorizes hard surfaces. Contec™ Sporicidin™ Disinfectant Solution is compatible with plastic, wood, glass, and metal surfaces.

  • Provides 100% kill of pathogenic vegetative organisms, including MRSA, VRE and Avian Influenza A Virus (H9N2 and H1N1)
  • Continuous residual activity up to 6 months
  • Compatible with plastics, wood, glass and metals
  • Non-staining, non-abrasive and non-corrosive
  • Alcohol-free
  • EPA and FDA registered
  • OSHA compliant
  • Lowest EPA toxicity category

Compliance

EPA and FDA registered, OSHA compliant, Lowest EPA toxicity category

Specifications

Product Type Disinfectant Solution
Form Liquid
pH 7.5 to 8.5
Product Line Sporicidin
Sterility Sterile
Appearance Clear
Certifications/Compliance FDA, OSHA, EPA
Composition 1.56% Buffered Phenol, 0.06% Sodium Phenate, 98.38% Inert Ingredients
For Use With (Application) Disinfecting hard surfaces in critical areas where USP <797> compliance has been mandated, such as ISO Class 8 Anterooms in hospitals and compounding pharmacies
Packaging Double bagged
Volume (English) 1 gal.
Volume (Metric) 3.8 L
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.