Description
- Highest sorbency in a cleanroom mop
- Constructed from 100% continuous filament polyester with unique two-yarn system for superior sorbency Designed for wet mopping in critical areas where maximum sorbency is required for applying cleaning and disinfectant solutions
- Mop is durable and launderable for multiple uses
- Designed for use with commercially available handles and bucket-wringer systems
- Autoclavable for sterile applications
- Constructed of 100% continuous filament polyester with unique two-yarn system for exceptional sorbency and mopping characteristics
- Can be used with commercially available hardware—no need for special handles
- Autoclavable for sterile applications
Specifications
Specifications
| Product Type | Cleanroom Mop |
| Autoclavable | Autoclavable |
| For Use With (Application) | Commercial Hardware |
| Certifications/Compliance | ISO Class 1 |
| Material | Continuous Filament Polyester |