Learn More
Contec™ VertiKlean™ Wall Washing Systems Sterile Mop Heads
BIOPHARMAGREENER_CHOICE
Catalog No. 19167026 Shop All Contec Inc. Products
Catalog No. 19-167-026 Supplier Contec™ Supplier No. TMVKMOPIR
Add to Cart
Add to Cart
Contec™ VertiKlean™ Wall Washing Systems Sterile Mop Heads and Covers are comprised of laminated polyester fabric and high sorbency polyester foam for excellent cleaning capacity and abrasion resistance.
Use with VertiKlean™ Wall Washing System which is designed for cleaning vertical surfaces. Contec™ VertiKlean™ Wall Washing Systems Sterile Mop Heads and Covers are comprised of laminated polyester fabric and high sorbency polyester foam for excellent cleaning capacity and abrasion resistance. A textured option for applications that require improved particle removal is available.
  • Lightweight, polyester, disposable
  • Ideal for applying disinfectants and wet cleaning vertical surfaces, ceilings, and walls in controlled environments; reaching awkward corners and ceilings with a complete range of motion
  • Absorbent foam carries solvents and disinfectants to the surface; polyester fabric offers excellent abrasion resistance and helps distribute the liquid uniformly on the surface being cleaned
  • Heads are laser cut to seal edges, further limiting potential contamination in the mopping process
  • Textured surface option is ideal for applications requiring additional scrubbing performance
  • Contain no cellulose; compatible with a wide range of chemicals, detergents, disinfectants, and cleaners
Specifications
Dimensions 12.5 x 14 x 1.25 in. (32 x 10 x 3 cm)
Sterility Sterile
Disposable Single-use
Color-coded No
Material Knitted Polyester, Polyester Foam