Learn More
Medchemexpress LLC HY-109084 5mg Medchemexpress, Conteltinib CAS:1384860-29-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1090845MG
Encompass_Preferred
Medchemexpress, HY-109084 5mg Conteltinib CAS:1384860-29-0 Conteltinib (CT-707) is a multi-kinase inhibitor targeting FAK, ALK, and Pyk2. Conteltinib (CT-707) exerts significant inhibitory effect on FAK with an IC50 of 1.6 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8669