Learn More
Medchemexpress LLC Continentalic acid | 19889-23-7 | 10 MG
SDP
Catalog No. 5000419870
Encompass_Preferred
Continentalic acid, isolated from Aralia continentalis, is a chemical compound primarily studied for its antibacterial properties. It is a white to off-white solid and is intended for research use only.
  • Demonstrates minimum inhibitory concentrations (MICs) of 8-16 μg/mL against S. aureus
  • Effective against Methicillin-susceptible Staphylococcus aureus (MSSA)
  • Effective against Methicillin-resistant Staphylococcus aureus (MRSA) standard strains
Catalog No. 50-004-19870 Supplier Medchemexpress LLC Supplier No. HYN690810MG
Continentalic acid, isolated from Aralia continentalis, is a chemical compound primarily studied for its antibacterial properties. It is a white to off-white solid and is intended for research use only.
  • Demonstrates minimum inhibitory concentrations (MICs) of 8-16 μg/mL against S. aureus
  • Effective against Methicillin-susceptible Staphylococcus aureus (MSSA)
  • Effective against Methicillin-resistant Staphylococcus aureus (MRSA) standard strains