Learn More
MSC Karcher Industrial Vacuum, 460V 3-Phase, 26.4 gal, 1 ea
Catalog No. 502818948
Encompass
This Karcher industrial vacuum is designed for continuous duty in fixed process applications. It utilizes completely enclosed and fan-cooled (TEFC) side-channel motors for high performance, allowing operators to work further from the unit. This vacuum is CSA approved and NFPA 652 compliant.
  • Easy-to-operate with simple knobs and easy-to-connect hoses.
  • Horizontal filter cleaning handle for easy manual filter cleaning.
  • Robust welded frame construction.
  • Tangential inlet helps prevent plugging with large debris.
  • Side-channel blower ensures low-wear operation.
  • M cartridge filter system for hazardous dust filtration.
  • Stainless steel collection container handles aggressive debris.
  • Optional HEPA filter available for critical filtration needs.
Catalog No. 50-281-8948 Supplier MSC Supplier No. 16617714
Add to Cart
Add to Cart
Request a Quote
This Karcher industrial vacuum is designed for continuous duty in fixed process applications. It utilizes completely enclosed and fan-cooled (TEFC) side-channel motors for high performance, allowing operators to work further from the unit. This vacuum is CSA approved and NFPA 652 compliant.
  • Easy-to-operate with simple knobs and easy-to-connect hoses.
  • Horizontal filter cleaning handle for easy manual filter cleaning.
  • Robust welded frame construction.
  • Tangential inlet helps prevent plugging with large debris.
  • Side-channel blower ensures low-wear operation.
  • M cartridge filter system for hazardous dust filtration.
  • Stainless steel collection container handles aggressive debris.
  • Optional HEPA filter available for critical filtration needs.