Learn More
Grainger 1 Gallon Jar, HDPE, Includes Closure, Wide, 24/pk
Catalog No. NC9485699
Encompass_Preferred
These round jars are suitable for liquid and solid specimen storage. Each features high-density polyethylene (HDPE) construction and offers resistance to formaldehyde and weak acids or bases. The wide mouth design is perfect for solids and powders.
Catalog No. NC9485699 Supplier Grainger Supplier No. 3UDH6
These round jars are suitable for liquid and solid specimen storage. Each features high-density polyethylene (HDPE) construction and offers resistance to formaldehyde and weak acids or bases. The wide mouth design is perfect for solids and powders.