Learn More
Glass Expansion Contour Flared End PVC Pump Tube 3tag, 72mm spacing, 1.30mm ID, Grey/Grey (PKT 6)
SDP
Supplier:  Glass Expansion 1.30GRY3BF
Encompass_Preferred
Contour Flared End PVC Pump Tube 3tag 1.30mm ID Gray/Gray (PKT 6).Tag Colors: Gray/GrayID mm: 1.30Wall Thickness mm: 0.86OD mm: 3.02Overall Length: 455mm (72mm spacing between tags)Number of Tubes Per Pack: 6Physical Properties: Thermoplastic, PVC based material with plasticizerMaterial: Made from Tygon R3603/R3607 or equivalent.Special Properties: Inexpensive all-around tubing for general laboratory applicationsChemical Compatibility: Excellent for aqueous solutions of the following acids and concentrations, up to 35% Nitric Acid, up to 37% Hydrochloric Acid, up to 25% Hydrofluoric Acid, up to 30% Hydrogen Peroxide.Limitations: Potential leaching of plasticizersGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6856