Learn More
Glass Expansion Contour Flared End PVC Pump Tube 3tag, 95mm spacing, 1.85mm ID, Green/Green, 95mm (PKT 6)
SDP
Supplier:  Glass Expansion 1.85GRN3B95F
Encompass_Preferred
Contour Flared End PVC Pump Tube 3tag (95mm spacing) 1.85mm ID Green/Green (PKT 6).Tag Colors: Green/GreenID mm: 1.85Wall Thickness mm: 0.86OD mm: 3.57Overall Length: 455mm (95mm spacing between tags)Number of Tubes Per Pack: 6Physical Properties: Thermoplastic, PVC based material with plasticizerMaterial: Made from Tygon R3603/R3607 or equivalent.Special Properties: Inexpensive all-around tubing for general laboratory applicationsChemical Compatibility: Excellent for aqueous solutions of the following acids and concentrations, up to 35% Nitric Acid, up to 37% Hydrochloric Acid, up to 25% Hydrofluoric Acid, up to 30% Hydrogen Peroxide.Limitations: Potential leaching of plasticizersGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. 50-190-6886