Learn More
Glass Expansion Contour Flared End PVC Pump Tube 3tag 1.02mm ID White/White (PKT 6)
SDP
Catalog No. NC2046953
Encompass_Preferred
Contour Flared End PVC Pump Tube 3tag 1.02mm ID White/White (PKT 6).Tag Colors: White/WhiteID mm: 1.02Wall Thickness mm: 0.86OD mm: 2.74Overall Length: 455mm (72mm spacing between tags)Number of Tubes Per Pack: 6Physical Properties: Thermoplastic, PVC based material with plasticizerMaterial: Made from Tygon R3603/R3607 or equivalent.Special Properties: Inexpensive all-around tubing for general laboratory applicationsChemical Compatibility: Excellent for aqueous solutions of the following acids and concentrations, up to 35% Nitric Acid, up to 37% Hydrochloric Acid, up to 25% Hydrofluoric Acid, up to 30% Hydrogen Peroxide.Limitations: Potential leaching of plasticizersGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001
Catalog No. NC2046953 Supplier Glass Expansion Supplier No. 1.02WHT3BF
Contour Flared End PVC Pump Tube 3tag 1.02mm ID White/White (PKT 6).Tag Colors: White/WhiteID mm: 1.02Wall Thickness mm: 0.86OD mm: 2.74Overall Length: 455mm (72mm spacing between tags)Number of Tubes Per Pack: 6Physical Properties: Thermoplastic, PVC based material with plasticizerMaterial: Made from Tygon R3603/R3607 or equivalent.Special Properties: Inexpensive all-around tubing for general laboratory applicationsChemical Compatibility: Excellent for aqueous solutions of the following acids and concentrations, up to 35% Nitric Acid, up to 37% Hydrochloric Acid, up to 25% Hydrofluoric Acid, up to 30% Hydrogen Peroxide.Limitations: Potential leaching of plasticizersGlass Expansion is ISO 9001:2015 certified by AB Certification, No. AU4501-9001