Learn More
Science First™ Contour Map Making Kit
SDPSCI_ED
Catalog No. S53005
Change view
Click to view available options
Quantity:
15
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S53005 Each
S53005B 15
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S53005 Supplier Science First™ Supplier No. 32200
Add to Cart
Add to Cart
Includes
  • Plastic sheet
  • Plastic mountain
  • Plastic box, 12L x 6W x 3 in.H
Learn how to read and make contour maps.
Easy to use—just cover the molded plastic model with water, mark mountain at each elevation, and then trace contour lines from above onto a transparent plastic sheet.
Specifications
Includes Plastic sheet, Plastic mountain, Plastic box
Product Type Contour Map Making Kit
Quantity Each