Learn More
MSC Smith Sprayer, 3 gal., Polyethylene, Commercial Grade, 1 ea
Catalog No. 502867100
Encompass
Smith Contractor MAX Sprayers are equipped with robust, professional-grade components in essential areas to deliver performance and value. These sprayers are engineered to achieve higher pressures and provide effective spray performance while using cost-effective poly components.
  • Versatile 3-gallon sprayer for heavy-duty commercial cleaning, sanitizing, and harsh chemical application.
  • Suitable for lawn and garden use.
  • Commercial-grade non-corrosive shut-off with comfortable grip, lock-on, and in-line filter.
  • Poly wand (21-inch) and 50-inch hose allow for precise material placement.
  • Viton seals provide long-term resistance to chemicals.
  • Integrated stability base with convenient wand storage.
  • Pressure release valve helps prevent chemical exposure.
  • Includes 64-inch carry strap and 7 versatile nozzles.
Catalog No. 50-286-7100 Supplier MSC Supplier No. 30302053
Smith Contractor MAX Sprayers are equipped with robust, professional-grade components in essential areas to deliver performance and value. These sprayers are engineered to achieve higher pressures and provide effective spray performance while using cost-effective poly components.
  • Versatile 3-gallon sprayer for heavy-duty commercial cleaning, sanitizing, and harsh chemical application.
  • Suitable for lawn and garden use.
  • Commercial-grade non-corrosive shut-off with comfortable grip, lock-on, and in-line filter.
  • Poly wand (21-inch) and 50-inch hose allow for precise material placement.
  • Viton seals provide long-term resistance to chemicals.
  • Integrated stability base with convenient wand storage.
  • Pressure release valve helps prevent chemical exposure.
  • Includes 64-inch carry strap and 7 versatile nozzles.