Learn More
MSC ORANGE-SOL Contractors Solvent, Flash Point >104.4 °C, Specific Gravity 0.8946
Catalog No. 502839258
Encompass
ORANGE-SOL De-Solv-it Contractors Solvent is a versatile solution designed for tough industrial cleaning tasks. It effectively removes a wide range of stubborn materials including asphalt, roofing tar, road tar, grease, oil, wet oil-based lacquers, paints, stains, adhesives, caulk, mastic, liquid foam insulation, silicone sealant, and water seal. This solvent is a combustible liquid and is biodegradable, making it a powerful yet environmentally conscious choice for contractors.
  • Effectively removes asphalt, roofing tar, and road tar
  • Cleans grease, oil, wet oil-based lacquers, paints, and stains
  • Dissolves adhesives, caulk, mastic, and liquid foam insulation
  • Removes silicone sealant and water seal
  • Biodegradable composition
Catalog No. 50-283-9258 Supplier MSC Supplier No. 73938508
ORANGE-SOL De-Solv-it Contractors Solvent is a versatile solution designed for tough industrial cleaning tasks. It effectively removes a wide range of stubborn materials including asphalt, roofing tar, road tar, grease, oil, wet oil-based lacquers, paints, stains, adhesives, caulk, mastic, liquid foam insulation, silicone sealant, and water seal. This solvent is a combustible liquid and is biodegradable, making it a powerful yet environmentally conscious choice for contractors.
  • Effectively removes asphalt, roofing tar, and road tar
  • Cleans grease, oil, wet oil-based lacquers, paints, and stains
  • Dissolves adhesives, caulk, mastic, and liquid foam insulation
  • Removes silicone sealant and water seal
  • Biodegradable composition