Learn More
Thomas Scientific CONTRAD 70 DETERG 1L UN1814-8 P2 FOR AIR
SDP
Catalog No. NC9831479
Encompass_Preferred
CONTRAD 70 Biodegradable Liquid Concentrate, 1L Bottle; Phosphate-free, Usable on glass, ceramics, plastics, rubber, ferrous metals, etc.
Catalog No. NC9831479 Supplier Thomas Scientific Supplier No. 2901C15
CONTRAD 70 Biodegradable Liquid Concentrate, 1L Bottle; Phosphate-free, Usable on glass, ceramics, plastics, rubber, ferrous metals, etc.