Learn More
Bachem H-Ile-Ile-OH
SDP
Supplier:  Bachem 4007632.0005
Encompass_Preferred
H-Ile-Ile-OH, 42537-99-5, C12H24N2O3, Mr 244.33, 5g. Contrary to similar non-polar dipeptides as Ile-Leu or Ile-Val, Ile-Ile crystallizes forming hydrophobic columns.
Catalog No. 50-194-6834